Learn More
Cayman Chemical Cholesteryl HomoLInolenAT 10mg
SDP
Catalog No. 502933509
Encompass_Preferred
A cholesterol ester; accumulates in the adrenal gland of rabbits fed a diet high in linolenic acid; levels are decreased and positively correlate with cognitive decline in HIV-infected humans
Catalog No. 50-293-3509 Supplier Cayman Chemical Supplier No. 2646910 MG
Add to Cart
Add to Cart
A cholesterol ester; accumulates in the adrenal gland of rabbits fed a diet high in linolenic acid; levels are decreased and positively correlate with cognitive decline in HIV-infected humans