Learn More
Cayman Chemical Cholesteryl Dihomo-γ-Linolenate, Purity: ≥98%, CAS: 7274-08-0, 25 mg
SDP
Catalog No. 502934011
Encompass_Preferred
A cholesterol ester; accumulates in the adrenal gland of rabbits fed a diet high in linolenic acid; levels are decreased and positively correlate with cognitive decline in HIV-infected humans
Catalog No. 50-293-4011 Supplier Cayman Chemical Supplier No. 2646925 MG
A cholesterol ester; accumulates in the adrenal gland of rabbits fed a diet high in linolenic acid; levels are decreased and positively correlate with cognitive decline in HIV-infected humans