Learn More
Cayman Chemical Cholesteryl Lignocerate, Purity: ≥98%, CAS: 73024-96-1, 10 mg
SDP
Catalog No. 502933458
Encompass_Preferred
A cholesterol ester that has been found in human meibum; can be hydrolyzed by cellular extracts from cultured human skin fibroblasts isolated from healthy individuals, but not patients with CESD or Wolman disease
Catalog No. 50-293-3458 Supplier Cayman Chemical Supplier No. 2647010 MG
A cholesterol ester that has been found in human meibum; can be hydrolyzed by cellular extracts from cultured human skin fibroblasts isolated from healthy individuals, but not patients with CESD or Wolman disease