Learn More
Cayman Chemical Cholesteryl LIgnocerate 5mg
SDP
Catalog No. 502935432
Encompass_Preferred
A cholesterol ester that has been found in human meibum; can be hydrolyzed by cellular extracts from cultured human skin fibroblasts isolated from healthy individuals, but not patients with CESD or Wolman disease
Catalog No. 50-293-5432 Supplier Cayman Chemical Supplier No. 264705 MG
Add to Cart
Add to Cart
A cholesterol ester that has been found in human meibum; can be hydrolyzed by cellular extracts from cultured human skin fibroblasts isolated from healthy individuals, but not patients with CESD or Wolman disease