Learn More
Cayman Chemical Cholesteryl LInoleATd11 100ug
SDP
Catalog No. 502917845
Encompass_Preferred
An internal standard for the quantification of cholesteryl linoleate by GC- or LC-MS
Catalog No. 50-291-7845 Supplier Cayman Chemical Supplier No. 37920100 UG
An internal standard for the quantification of cholesteryl linoleate by GC- or LC-MS