Learn More
Cayman Chemical Cholesteryl LInoleate 100mg
SDP
Catalog No. 502964134
Encompass_Preferred
A pro-atherogenic cholesterol ester; has been found in the plasma membrane of macrophages as a component of LDL; levels are increased in apoB-lipoproteins and positively correlated with hepatic cholesterol levels and the number of atherosclerotic lesions in transgenic mice overexpressing human ABCA1; lipoprotein levels are increased and associated with lipid radical formation in patients with β-thalassemia
Catalog No. 50-296-4134 Supplier Cayman Chemical Supplier No. 22597100 MG
Add to Cart
Add to Cart
A pro-atherogenic cholesterol ester; has been found in the plasma membrane of macrophages as a component of LDL; levels are increased in apoB-lipoproteins and positively correlated with hepatic cholesterol levels and the number of atherosclerotic lesions in transgenic mice overexpressing human ABCA1; lipoprotein levels are increased and associated with lipid radical formation in patients with β-thalassemia