Learn More
Cayman Chemical Cholesteryl Linoleate, Purity: ≥98%, CAS: 604-33-1, 250 mg
SDP
Catalog No. 502964072
Encompass_Preferred
A pro-atherogenic cholesterol ester; has been found in the plasma membrane of macrophages as a component of LDL; levels are increased in apoB-lipoproteins and positively correlated with hepatic cholesterol levels and the number of atherosclerotic lesions in transgenic mice overexpressing human ABCA1; lipoprotein levels are increased and associated with lipid radical formation in patients with β-thalassemia
Catalog No. 50-296-4072 Supplier Cayman Chemical Supplier No. 22597250 MG
A pro-atherogenic cholesterol ester; has been found in the plasma membrane of macrophages as a component of LDL; levels are increased in apoB-lipoproteins and positively correlated with hepatic cholesterol levels and the number of atherosclerotic lesions in transgenic mice overexpressing human ABCA1; lipoprotein levels are increased and associated with lipid radical formation in patients with β-thalassemia