Learn More
Cayman Chemical Cholesteryl Linoleate Hydrope
SDP
Catalog No. 502945166
Encompass_Preferred
Cholesteryl linoleate hydroperoxides are derived from the autoxidation of cholesteryl linoleate and contain a mixture of racemic 9- and 13-HpODE cholesteryl esters. Oxidative modification of LDL is suggested to play an important role in atherosclerosis. (±)9- and (±)13-HODE cholesteryl esters were originally extracted from atherosclerotic lesions and shown to be produced by Cu2+-catalyzed oxidation of LDL.{2227,2228} Later studies determined that 15-LO, from rabbit reticulocytes and activated human monocytes, oxygenates cholesteryl linoleate to both 9- and 13-hydroperoxy linoleate cholesteryl esters.{1126,2412} Cholesteryl ester hydroperoxides may be transferred from LDL to HDL, reduced to the corresponding hydroxides, and cleared via the liver.{6527,3832}
Catalog No. 50-294-5166 Supplier Cayman Chemical Supplier No. 480011 MG
Add to Cart
Add to Cart
Cholesteryl linoleate hydroperoxides are derived from the autoxidation of cholesteryl linoleate and contain a mixture of racemic 9- and 13-HpODE cholesteryl esters. Oxidative modification of LDL is suggested to play an important role in atherosclerosis. (±)9- and (±)13-HODE cholesteryl esters were originally extracted from atherosclerotic lesions and shown to be produced by Cu2+-catalyzed oxidation of LDL.{2227,2228} Later studies determined that 15-LO, from rabbit reticulocytes and activated human monocytes, oxygenates cholesteryl linoleate to both 9- and 13-hydroperoxy linoleate cholesteryl esters.{1126,2412} Cholesteryl ester hydroperoxides may be transferred from LDL to HDL, reduced to the corresponding hydroxides, and cleared via the liver.{6527,3832}