Learn More
Cayman Chemical Cholesteryl Linoleate Hydroperoxides, Purity: ≥98% hydroperoxide content, 5 mg
SDP
Catalog No. 502944995
Encompass_Preferred
Cholesteryl linoleate hydroperoxides are derived from the autoxidation of cholesteryl linoleate and contain a mixture of racemic 9- and 13-HpODE cholesteryl esters. Oxidative modification of LDL is suggested to play an important role in atherosclerosis. (±)9- and (±)13-HODE cholesteryl esters were originally extracted from atherosclerotic lesions and shown to be produced by Cu2+-catalyzed oxidation of LDL.{2227,2228} Later studies determined that 15-LO, from rabbit reticulocytes and activated human monocytes, oxygenates cholesteryl linoleate to both 9- and 13-hydroperoxy linoleate cholesteryl esters.{1126,2412} Cholesteryl ester hydroperoxides may be transferred from LDL to HDL, reduced to the corresponding hydroxides, and cleared via the liver.{6527,3832}
Catalog No. 50-294-4995 Supplier Cayman Chemical Supplier No. 480015 MG
Cholesteryl linoleate hydroperoxides are derived from the autoxidation of cholesteryl linoleate and contain a mixture of racemic 9- and 13-HpODE cholesteryl esters. Oxidative modification of LDL is suggested to play an important role in atherosclerosis. (±)9- and (±)13-HODE cholesteryl esters were originally extracted from atherosclerotic lesions and shown to be produced by Cu2+-catalyzed oxidation of LDL.{2227,2228} Later studies determined that 15-LO, from rabbit reticulocytes and activated human monocytes, oxygenates cholesteryl linoleate to both 9- and 13-hydroperoxy linoleate cholesteryl esters.{1126,2412} Cholesteryl ester hydroperoxides may be transferred from LDL to HDL, reduced to the corresponding hydroxides, and cleared via the liver.{6527,3832}