Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesteryl Linoleate , MP Biomedicals, LLC

Catalog No. MP210044491 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210044491 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210044491 Supplier MP Biomedicals, Inc Supplier No. 0210044491
Add to Cart
Add to Cart

Cholesteryl Linoleate ≥99% is used as an HPLC standard in the analysis of rat lipoproteins.

  • HPLC standard in the analysis of rat liver lipoproteins
  • The most abundant cholesteryl ester in low-density lipoprotein (LDL)
  • Major cholesterol ester found associated with the neutral core of low-density lipoprotein
  • Receptor-LDL complexes are taken up by lysosomes and hydrolyzed to release cholesterol from the esters
  • Enzyme acid cholesteryl ester hydrolase is responsible for the hydrolysis of cholesteryl esters
  • A defective enzyme can result in the formation of atherosclerotic lesions in humans
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 604-33-1
Molecular Formula C45H76O2
Molecular Weight (g/mol) 649.10
InChI Key NAACPBBQTFFYQB-LJAITQKLSA-N
Synonym cholesteryl linoleate, 18:2 cholesteryl ester, cholesterol linoleate, linoleic acid cholesterol ester, cholest-5-en-3beta-yl z,z-octadeca-9,12-dienoate, cholesteryl 9z,12z-octadecadienoate, z,z-3beta-cholest-5-en-3-ol 9,12-octadecadienoate, cholest-5-en-3beta-yl 9z,12z-octadecadienoate, 3beta-cholest-5-en-3-ol, z,z-9,12-octadecadienoate, 3beta-cholest-5-en-3-ol, 9z,12z-9,12-octadecadienoate
PubChem CID 131632555

Specifications

Melting Point 39°C to 41°C (literature)
Color Colorless
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.