Learn More
Cayman Chemical Cholesteryl MyrIstate 500mg
SDP
Catalog No. 502933587
Encompass_Preferred
A cholesterol ester; levels are increased in the serum, liver, and aorta in rabbits fed a high-cholesterol diet and positively correlate with development of NAFLD; plasma levels positively correlate with the presence of unstable atherosclerotic plaques in coronary artery disease; has been used in the composition of lipid nanoparticles as drug carrier systems for drugs with low water solubility
Catalog No. 50-293-3587 Supplier Cayman Chemical Supplier No. 26439500 MG
A cholesterol ester; levels are increased in the serum, liver, and aorta in rabbits fed a high-cholesterol diet and positively correlate with development of NAFLD; plasma levels positively correlate with the presence of unstable atherosclerotic plaques in coronary artery disease; has been used in the composition of lipid nanoparticles as drug carrier systems for drugs with low water solubility