Learn More
Cayman Chemical Cholesteryl Nervonate, Purity: ≥98%, CAS: 60758-73-8, 10 mg
SDP
Catalog No. 502933459
Encompass_Preferred
A cholesterol ester; has been found in human meibum; has been used in the generation of liposomes
Catalog No. 50-293-3459 Supplier Cayman Chemical Supplier No. 2647110 MG
A cholesterol ester; has been found in human meibum; has been used in the generation of liposomes