Learn More
Cayman Chemical Cholesteryl Nervonate, Purity: ≥98%, CAS: 60758-73-8, 25 mg
SDP
Catalog No. 502934567
Encompass_Preferred
A cholesterol ester; has been found in human meibum; has been used in the generation of liposomes
Catalog No. 50-293-4567 Supplier Cayman Chemical Supplier No. 2647125 MG
A cholesterol ester; has been found in human meibum; has been used in the generation of liposomes