Learn More
Cayman Chemical Cholesteryl nrvonate 50mg
SDP
Catalog No. 502933460
Encompass_Preferred
A cholesterol ester; has been found in human meibum; has been used in the generation of liposomes
Catalog No. 50-293-3460 Supplier Cayman Chemical Supplier No. 2647150 MG
A cholesterol ester; has been found in human meibum; has been used in the generation of liposomes