Learn More
Cayman Chemical Cholesteryl Oleate, Purity: ≥95%, CAS: 303-43-5, 250 mg
SDP
Catalog No. 502933461
Encompass_Preferred
A cholesterol ester; has been used in the generation of nanoparticles and liposomes for drug delivery
Catalog No. 50-293-3461 Supplier Cayman Chemical Supplier No. 26472250 MG
A cholesterol ester; has been used in the generation of nanoparticles and liposomes for drug delivery