Learn More
Medchemexpress LLC Cholesteryl oleate | 303-43-5 | 99.0% | 651.10 | 100 MG
SDP
Catalog No. 5000364710
Encompass_Preferred
Cholesteryl oleate is an ester compound formed from Cholesterol and Oleic acid. It is involved in lipid transport, storage, and cell membrane formation in living organisms. This compound may serve as a potential biomarker for prostate cancer. Cholesteryl oleate can also be used to prepare cationic solid lipid nanoparticles (SLNs) for efficient gene silencing. The enrichment of Cholesteryl oleate may be a key factor in promoting atherosclerosis.
  • Involved in lipid transport, storage, and cell membrane formation
  • Potential biomarker for prostate cancer
  • Can be used to prepare cationic solid lipid nanoparticles for efficient gene silencing
  • Enrichment may be a key factor in promoting atherosclerosis
Catalog No. 50-003-64710 Supplier Medchemexpress LLC Supplier No. 100MGHY113217
Cholesteryl oleate is an ester compound formed from Cholesterol and Oleic acid. It is involved in lipid transport, storage, and cell membrane formation in living organisms. This compound may serve as a potential biomarker for prostate cancer. Cholesteryl oleate can also be used to prepare cationic solid lipid nanoparticles (SLNs) for efficient gene silencing. The enrichment of Cholesteryl oleate may be a key factor in promoting atherosclerosis.
  • Involved in lipid transport, storage, and cell membrane formation
  • Potential biomarker for prostate cancer
  • Can be used to prepare cationic solid lipid nanoparticles for efficient gene silencing
  • Enrichment may be a key factor in promoting atherosclerosis