Learn More
Medchemexpress LLC Cholesteryl oleate | 303-43-5 | 99.0% | 651.10 | 50 MG
SDP
Catalog No. 5000437231
Encompass_Preferred
Cholesteryl oleate is an ester compound formed from Cholesterol and Oleic acid, involved in lipid transport, storage, and cell membrane formation in living organisms. It may serve as a potential biomarker for prostate cancer and can be used to prepare cationic solid lipid nanoparticles (SLNs) for efficient gene silencing.
  • Involved in lipid transport, storage, and cell membrane formation in living organisms.
  • May serve as a potential biomarker for prostate cancer.
  • Can be used to prepare cationic solid lipid nanoparticles (SLNs) for efficient gene silencing.
  • SLNs prepared with cholesteryl oleate can efficiently deliver DNA or siRNA to cells without cytotoxicity and have good transfection efficiency.
Catalog No. 50-004-37231 Supplier Medchemexpress LLC Supplier No. HY11321750MG
Cholesteryl oleate is an ester compound formed from Cholesterol and Oleic acid, involved in lipid transport, storage, and cell membrane formation in living organisms. It may serve as a potential biomarker for prostate cancer and can be used to prepare cationic solid lipid nanoparticles (SLNs) for efficient gene silencing.
  • Involved in lipid transport, storage, and cell membrane formation in living organisms.
  • May serve as a potential biomarker for prostate cancer.
  • Can be used to prepare cationic solid lipid nanoparticles (SLNs) for efficient gene silencing.
  • SLNs prepared with cholesteryl oleate can efficiently deliver DNA or siRNA to cells without cytotoxicity and have good transfection efficiency.