Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesteryl Oleate 99+%, Thermo Scientific™

Catalog No. AC165840050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC165840050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC165840050 Supplier Thermo Scientific Chemicals Supplier No. 165840050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303-43-5
Molecular Formula C45H78O2
Molecular Weight (g/mol) 651.12
MDL Number MFCD00003645,MFCD00065898
InChI Key RJECHNNFRHZQKU-RMUVNZEASA-N
Synonym cholest-5-en-3-yl 9-octadecenoate, cholest-5-en-3-yl octadec-9-enoate
PubChem CID 54269525
IUPAC Name (1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl (9Z)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C

Specifications

Melting Point 42°C to 48°C
Color White to Yellow
Loss on Drying 0.2% max. (105°C, 2 hrs, 1mm Hg)
Infrared Spectrum Authentic
Quantity 5 g
Specific Rotation −23.5° to −25.5° (20°C, 589nm) (c=2, CHCl3)
Formula Weight 651.1
Percent Purity ≥99.0%
Physical Form Liquid or Solid
Chemical Name or Material Cholesteryl oleate, 99+%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.