Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cholesteryl oleate-d7 | 1416275-35-8 | 99% | 658.14 | 1 MG
SDP

Catalog No. 5000437242
Encompass_Preferred

Cholesteryl oleate-d7 | 1416275-35-8 | 99% | 658.14 | 1 MG

Catalog No. 50-004-37242 Supplier Medchemexpress LLC Supplier No. HY113217S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cholesteryl oleate-d7 | 1416275-35-8 | 99% | 658.14 | 1 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.