Learn More
Cayman Chemical Cholesteryl Palmitate, Purity: ≥95%, CAS: 601-34-3, 1 g
SDP
Catalog No. 502934483
Encompass_Preferred
A cholesterol ester; a major cholesterol ester found in human meibum; has been used to form a stable tear film to study amphiphilic block copolymers as surfactants for dry eyes; levels in amniotic fluid are decreased in pregnancies in which infants later developed respiratory distress syndrome; levels in amniotic fluid are decreased in patients with well-controlled diabetes; levels are increased in BALF from patients with chronic interstitial pneumonia
Catalog No. 50-293-4483 Supplier Cayman Chemical Supplier No. 264731 G
A cholesterol ester; a major cholesterol ester found in human meibum; has been used to form a stable tear film to study amphiphilic block copolymers as surfactants for dry eyes; levels in amniotic fluid are decreased in pregnancies in which infants later developed respiratory distress syndrome; levels in amniotic fluid are decreased in patients with well-controlled diabetes; levels are increased in BALF from patients with chronic interstitial pneumonia