Learn More
Cayman Chemical Cholesteryl Palmitoleate, Purity: ≥95%, CAS: 16711-66-3, 250 mg
SDP
Catalog No. 502933632
Encompass_Preferred
A cholesterol ester; plasma levels are increased in ApoE-/- mice exposed to cigarette smoke and in pediatric patients with biliary atresia; has been used as a standard for the identification of cholesterol esters in human meibomian gland secretions
Catalog No. 50-293-3632 Supplier Cayman Chemical Supplier No. 26474250 MG
A cholesterol ester; plasma levels are increased in ApoE-/- mice exposed to cigarette smoke and in pediatric patients with biliary atresia; has been used as a standard for the identification of cholesterol esters in human meibomian gland secretions