Learn More
Cayman Chemical Cholesteryl PalmItoleate 50mg
SDP
Catalog No. 502934017
Encompass_Preferred
A cholesterol ester; plasma levels are increased in ApoE-/- mice exposed to cigarette smoke and in pediatric patients with biliary atresia; has been used as a standard for the identification of cholesterol esters in human meibomian gland secretions
Catalog No. 50-293-4017 Supplier Cayman Chemical Supplier No. 2647450 MG
A cholesterol ester; plasma levels are increased in ApoE-/- mice exposed to cigarette smoke and in pediatric patients with biliary atresia; has been used as a standard for the identification of cholesterol esters in human meibomian gland secretions