Learn More
Medchemexpress LLC Cholesteryl polyethylene glycol | 69068-97-9 | MFCD00151772 | <1.0% | 1055.60 g/mol | C68H110O8 | 50 MG
SDP
Catalog No. 5000283588
Encompass_Preferred
Cholesterol-PEG 600 is a cholesterol-polyethylene glycol conjugate that combines a hydrophobic cholesterol moiety with a PEG 600 chain to produce an amphiphilic reagent. It is supplied as an oil-phase biochemical for use in formulation, conjugation, and delivery research where membrane anchoring or increased solubility is required.
  • Provides a hydrophobic cholesterol moiety for membrane anchoring.
  • PEG 600 chain improves aqueous solubility and biocompatibility.
  • Enables conjugation to oligonucleotides, peptides, and other biomolecules.
  • Suitable for liposome, micelle, and nanoparticle surface modification.
  • Off-white to light yellow oil form for liquid-phase handling.
Catalog No. 50-002-83588 Supplier Medchemexpress LLC Supplier No. HYW12755850MG
Cholesterol-PEG 600 is a cholesterol-polyethylene glycol conjugate that combines a hydrophobic cholesterol moiety with a PEG 600 chain to produce an amphiphilic reagent. It is supplied as an oil-phase biochemical for use in formulation, conjugation, and delivery research where membrane anchoring or increased solubility is required.
  • Provides a hydrophobic cholesterol moiety for membrane anchoring.
  • PEG 600 chain improves aqueous solubility and biocompatibility.
  • Enables conjugation to oligonucleotides, peptides, and other biomolecules.
  • Suitable for liposome, micelle, and nanoparticle surface modification.
  • Off-white to light yellow oil form for liquid-phase handling.