Learn More
Medchemexpress LLC Cholesteryl stearate | 35602-69-8 | 98.0% | 653.13 | 5 G
SDP
Catalog No. 5000451773
Encompass_Preferred
Cholesteryl stearate is an ester compound found in both animal and plant sources. It is formed from cholesteryl alcohol and stearic acid, creating a stable compound used in various industrial and biomedical applications. It functions as a lubricant, thickener, and emollient in cosmetic products like lotions and creams, and is also used in the production of certain polymers and plastics. For research use only, it serves as a biochemical reagent for life science research.
  • Stable compound
  • Used as a lubricant, thickener, and emollient in cosmetic formulations
  • Utilized in the production of certain types of polymers and plastics
  • Suitable as a biochemical reagent for life science-related research
Catalog No. 50-004-51773 Supplier Medchemexpress LLC Supplier No. HYW0106955G
Cholesteryl stearate is an ester compound found in both animal and plant sources. It is formed from cholesteryl alcohol and stearic acid, creating a stable compound used in various industrial and biomedical applications. It functions as a lubricant, thickener, and emollient in cosmetic products like lotions and creams, and is also used in the production of certain polymers and plastics. For research use only, it serves as a biochemical reagent for life science research.
  • Stable compound
  • Used as a lubricant, thickener, and emollient in cosmetic formulations
  • Utilized in the production of certain types of polymers and plastics
  • Suitable as a biochemical reagent for life science-related research