Learn More
Cayman Chemical Cholesteryl Stearate, Purity: ≥95%, CAS: 35602-69-8, 500 mg
SDP
Catalog No. 502934484
Encompass_Preferred
A cholesterol ester; has been used as an internal standard for the quantification of sterol esters in various plant tissues by Q-TOF MS; levels are decreased in coronary arteries with atherosclerotic plaques, isolated fatty streaks, and isolated atherosclerotic plaques
Catalog No. 50-293-4484 Supplier Cayman Chemical Supplier No. 26475500 MG
A cholesterol ester; has been used as an internal standard for the quantification of sterol esters in various plant tissues by Q-TOF MS; levels are decreased in coronary arteries with atherosclerotic plaques, isolated fatty streaks, and isolated atherosclerotic plaques