Learn More
Cayman Chemical CholesTRol Uptake CelBased 1ea
SDP
Catalog No. 502894372
Encompass_Preferred
Maintaining body cholesterol homeostasis is critical for normal physiological functions and is achieved by a highly regulated balance of de novo synthesis, dietary cholesterol absorption, biliary clearance, and excretion. The mechanisms controlling cholesterol absorption are important to understand for the prevention and treatment of hypercholesterolemia. Cayman’s Cholesterol Uptake Cell-Based Assay Kit provides a convenient tool for studying cholesterol absorption modulators. The kit employs NBD Cholesterol, a fluorescently tagged cholesterol, as a probe for the detection of cholesterol taken up by cultured cells. U-18666A, a cholesterol intracellular trafficking inhibitor, is included as a positive control.
Catalog No. 50-289-4372 Supplier Cayman Chemical Supplier No. 6004401 EA
Maintaining body cholesterol homeostasis is critical for normal physiological functions and is achieved by a highly regulated balance of de novo synthesis, dietary cholesterol absorption, biliary clearance, and excretion. The mechanisms controlling cholesterol absorption are important to understand for the prevention and treatment of hypercholesterolemia. Cayman’s Cholesterol Uptake Cell-Based Assay Kit provides a convenient tool for studying cholesterol absorption modulators. The kit employs NBD Cholesterol, a fluorescently tagged cholesterol, as a probe for the detection of cholesterol taken up by cultured cells. U-18666A, a cholesterol intracellular trafficking inhibitor, is included as a positive control.