Learn More
Cayman Chemical Cholesterol-PEG(2000)-Biotin, Purity: ≥90%, 10 mg
SDP
Catalog No. 502918494
Encompass_Preferred
A biotinylated and PEGylated form of cholesterol; has been used as a lipid anchor to immobilize vesicles, liposomes, viral particles, bacteria, or yeast to microfluidic chips; increases phycoerythrin-conjugated streptavidin density on the surface of mesenchymal stem cells greater than sulfo-NHS-LC-biotin at 10 µM
Catalog No. 50-291-8494 Supplier Cayman Chemical Supplier No. 3903610 MG
A biotinylated and PEGylated form of cholesterol; has been used as a lipid anchor to immobilize vesicles, liposomes, viral particles, bacteria, or yeast to microfluidic chips; increases phycoerythrin-conjugated streptavidin density on the surface of mesenchymal stem cells greater than sulfo-NHS-LC-biotin at 10 µM