Learn More
Medchemexpress LLC Cholestyramine resin | 11041-12-6 | MFCD00130784 | 13.0-17.0% | Not applicable (polymeric, no defined molecular weight) | C27H47N | 25 G
SDP
Supplier:  Medchemexpress LLC HY10408125G
Encompass_Preferred
Cholestyramine is a polymeric bile acid-binding anion-exchange resin supplied as a dry powder for laboratory research. It binds intestinal bile acids, reducing their reabsorption and thereby increasing bile acid synthesis from cholesterol. The material is an insoluble styrene/divinylbenzene-based resin with quaternary ammonium functional groups and variable molecular weight.
  • Binds intestinal bile acids effectively.
  • Reduces bile acid reabsorption and increases bile acid synthesis from cholesterol.
  • Insoluble polymeric resin suitable for preparation of aqueous suspensions.
  • Anion-exchange resin containing quaternary ammonium functional groups.
  • Supplied as a dry powder for laboratory and research use.
  • Used in studies of cholesterol and bile acid metabolism.
Catalog No. 50-002-42207