Learn More
Description
Cholestyramine Resin USP is a fine powder white to buff in color that is hygroscopic. It is odorless or has not more than a slight amine-like odor.
Alternative name(s): cholestyramine,
Specifications
Specifications
| Chemical Name or Material | Cholestyramine Resin USP |
| Packaging | Jar, 100mL, White, HDPE, Round, 38-400, TE Pano Neck |
| CAS | 11041-12-6 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | Not applicable |
| Quantity | 25 g |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
