Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholestyramine Resin, USP, Spectrum™ Chemical
SDP

Catalog No. 18601659 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-659 25 g Amber Glass Bottle
18-601-660 100 g Amber Glass Bottle
18-601-661 500 g Amber Glass Bottle
18-601-662 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-659 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C173425GM
Add to Cart
Add to Cart

C1734, 11041-12-6

Spectrum™ Chemical Cholestyramine Resin, USP is a cholesterol lowering agent. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 11041-12-6
Molecular Formula C21H30ClN
Molecular Weight (g/mol) 331.93
InChI Key KNDHRUPPBXRELB-UHFFFAOYNA-M
IUPAC Name 4-[3-(4-ethylphenyl)butyl]-N,N,N-trimethylanilinium chloride
SMILES [Cl-].CCC1=CC=C(C=C1)C(C)CCC1=CC=C(C=C1)[N+](C)(C)C

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 12%
Packaging Amber Glass Bottle
Quantity 25 g
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.