Learn More
Apexbio Technology LLC Cholic acid 81-25-4 10mM (in 1mL Ethanol)
SDP
Catalog No. 5000063541
Encompass_Preferred
Cholic acid (CAS 81-25-4) is a naturally occurring bile acid characterized by amphipathic properties enabling it to function effectively as an ionic mild detergent under non-denaturing conditions This molecule interacts with lipid membranes altering permeability and facilitating the disruption of protein-lipid interactions without denaturing integral membrane proteins Due to this mechanism cholic acid is widely utilized in biochemical research protocols particularly for the solubilization extraction and purification of biologically active membrane proteins and receptor complexes
Catalog No. 50-000-63541 Supplier Apexbio Technology LLC Supplier No. N16805.1
Cholic acid (CAS 81-25-4) is a naturally occurring bile acid characterized by amphipathic properties enabling it to function effectively as an ionic mild detergent under non-denaturing conditions This molecule interacts with lipid membranes altering permeability and facilitating the disruption of protein-lipid interactions without denaturing integral membrane proteins Due to this mechanism cholic acid is widely utilized in biochemical research protocols particularly for the solubilization extraction and purification of biologically active membrane proteins and receptor complexes