Learn More
Cayman Chemical CholIc AcId-24-13C 10mg
SDP
Catalog No. 502943578
Encompass_Preferred
An internal standard for the quantification of cholic acid by GC- or LC-MS
Catalog No. 50-294-3578 Supplier Cayman Chemical Supplier No. 4028810 MG
Add to Cart
Add to Cart
An internal standard for the quantification of cholic acid by GC- or LC-MS