Learn More
Cayman Chemical Cholic Acid-24-13C, Purity: ≥98%, CAS: 52886-36-9, 1 mg
SDP
Catalog No. 502945902
Encompass_Preferred
An internal standard for the quantification of cholic acid by GC- or LC-MS
Catalog No. 50-294-5902 Supplier Cayman Chemical Supplier No. 402881 MG
An internal standard for the quantification of cholic acid by GC- or LC-MS