Learn More
Cayman Chemical CholIc AcId 250g
SDP
Catalog No. 502983514
Encompass_Preferred
A primary bile acid; induces C. difficile colony formation in an agar dilution assay at 0.1% w/v; dietary administration increases serum cholesterol levels, biliary phospholipid secretion, and fecal DCA levels in rats at 0.4% w/w
Catalog No. 50-298-3514 Supplier Cayman Chemical Supplier No. 20250250 G
A primary bile acid; induces C. difficile colony formation in an agar dilution assay at 0.1% w/v; dietary administration increases serum cholesterol levels, biliary phospholipid secretion, and fecal DCA levels in rats at 0.4% w/w