Learn More
Medchemexpress LLC Cholic acid 3-O-glucuronide disodium | 59274-69-0 | 628.66 | C30H46Na2O11 | 1 MG
SDP
Catalog No. 5000284865
Encompass_Preferred
Cholic acid 3-O-glucuronide disodium is the disodium salt of the 3-O-glucuronide metabolite of cholic acid. It functions as a farnesoid X receptor (FXR) agonist and is used in research on bile acid metabolism and detoxification. Supplied in small milligram quantities, QC reports and a Certificate of Analysis are available on request; intended for laboratory research use only.
  • Acts as an FXR agonist for bile acid metabolism studies.
  • Supplied as a disodium salt with formula C30H46Na2O11 and molecular weight 628.66.
  • Available in small packaging sizes suitable for analytical assays.
  • Quality control documentation and CoA available on request.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-84865 Supplier Medchemexpress LLC Supplier No. HYW5874511MG
Cholic acid 3-O-glucuronide disodium is the disodium salt of the 3-O-glucuronide metabolite of cholic acid. It functions as a farnesoid X receptor (FXR) agonist and is used in research on bile acid metabolism and detoxification. Supplied in small milligram quantities, QC reports and a Certificate of Analysis are available on request; intended for laboratory research use only.
  • Acts as an FXR agonist for bile acid metabolism studies.
  • Supplied as a disodium salt with formula C30H46Na2O11 and molecular weight 628.66.
  • Available in small packaging sizes suitable for analytical assays.
  • Quality control documentation and CoA available on request.
  • For research use only; not for human or clinical use.