Learn More
Cayman Chemical CholIc AcId 7-sulfate 10mg
SDP
Catalog No. 502893176
Encompass_Preferred
A metabolite of cholic acid; produced by conjugation of a sulfate group with the hydroxy group at position 7 of cholic acid in the liver and gut; fecal levels increased in male mice fed a diet supplemented with cholic acid or chenodeoxycholic acid
Catalog No. 50-289-3176 Supplier Cayman Chemical Supplier No. 900253210 MG
A metabolite of cholic acid; produced by conjugation of a sulfate group with the hydroxy group at position 7 of cholic acid in the liver and gut; fecal levels increased in male mice fed a diet supplemented with cholic acid or chenodeoxycholic acid