Learn More
Medchemexpress LLC Cholic acid 7-sulfate | 60320-05-0 | 99.9% | 10 MG
SDP
Catalog No. 5000343009
Encompass_Preferred
Cholic acid 7-sulfate (7-Sulfocholic acid) is a selective agonist targeting TGR5 (EC50=0.17 μM) and a ligand for MHC class I-related protein (MR1). It is primarily used in the research of diabetes and MAIT cell-related immune regulation.
  • Gut-restricted TGR5 agonist
  • Induces GLP-1 secretion and improves glucose tolerance
  • Promotes survival of mucosal-associated invariant T cells (MAIT)
  • Affects MAIT cell development and function
Catalog No. 50-003-43009 Supplier Medchemexpress LLC Supplier No. HY12685510MG
Cholic acid 7-sulfate (7-Sulfocholic acid) is a selective agonist targeting TGR5 (EC50=0.17 μM) and a ligand for MHC class I-related protein (MR1). It is primarily used in the research of diabetes and MAIT cell-related immune regulation.
  • Gut-restricted TGR5 agonist
  • Induces GLP-1 secretion and improves glucose tolerance
  • Promotes survival of mucosal-associated invariant T cells (MAIT)
  • Affects MAIT cell development and function