Learn More
Selleck Chemical LLC Cholic acid S3742-1g
SDP
Catalog No. 503109983
Encompass_Preferred
Cholic acid is a naturally occurring bile acid that is insoluble in water and is used to treat patients with genetic deficiencies in the synthesis of bile acids
Catalog No. 50-310-9983 Supplier Selleck Chemical LLC Supplier No. S37421G
Cholic acid is a naturally occurring bile acid that is insoluble in water and is used to treat patients with genetic deficiencies in the synthesis of bile acids