Learn More
American Radiolabeled Chemicals Inc CHOLIC ACID-CARB-14C 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARC 026050UC
Encompass_Preferred
Cholic acid-carboxyl-14C 50uCi in ethanol, 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 408.6
Catalog No. 50-901-05684