Learn More
Cayman Chemical Cholic Acid-d4, Purity: ≥95%, CAS: 116380-66-6, 10 mg
SDP
Catalog No. 502985103
Encompass_Preferred
An internal standard for the quantification of cholic acid by GC- or LC-MS
Catalog No. 50-298-5103 Supplier Cayman Chemical Supplier No. 2084910 MG
An internal standard for the quantification of cholic acid by GC- or LC-MS