Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cholic acid-d4 | 116380-66-6 | MFCD00209701 | 99.9% | 412.60 | C24H36D4O5 | 10 MG
SDP

Catalog No. 5000295536
Encompass_Preferred

Cholic acid-d4 is a deuterium-labeled form of cholic acid used as an internal standard and tracer in analytical workflows. Supplied as a white to off-white solid with high isotopic enrichment and purity, it is intended for research use and optimized for quantitative analysis by NMR, GC-MS, and LC-MS.

  • High purity (~99.9%) suitable for quantitative assays.
  • Molecular formula C24H36D4O5, molecular weight 412.60.
  • Functions as a tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Packaged as a 10 mg vial for analytical use.

Catalog No. 50-002-95536 Supplier Medchemexpress LLC Supplier No. HYN0324S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cholic acid-d4 is a deuterium-labeled form of cholic acid used as an internal standard and tracer in analytical workflows. Supplied as a white to off-white solid with high isotopic enrichment and purity, it is intended for research use and optimized for quantitative analysis by NMR, GC-MS, and LC-MS.

  • High purity (~99.9%) suitable for quantitative assays.
  • Molecular formula C24H36D4O5, molecular weight 412.60.
  • Functions as a tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Packaged as a 10 mg vial for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.