Learn More
Cayman Chemical CholIc AcId-d4 1mg
SDP
Catalog No. 502982254
Encompass_Preferred
An internal standard for the quantification of cholic acid by GC- or LC-MS
Catalog No. 50-298-2254 Supplier Cayman Chemical Supplier No. 208491 MG
An internal standard for the quantification of cholic acid by GC- or LC-MS