Learn More
Cayman Chemical Cholic Acid-d4, Purity: ≥95%, CAS: 116380-66-6, 5 mg
SDP
Catalog No. 502982642
Encompass_Preferred
An internal standard for the quantification of cholic acid by GC- or LC-MS
Catalog No. 50-298-2642 Supplier Cayman Chemical Supplier No. 208495 MG
An internal standard for the quantification of cholic acid by GC- or LC-MS