Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cholic acid-d5 | 53007-09-3 | MFCD08702879 | 98.0% | 413.60 | C24H35D5O5 | 10 MG
SDP

Catalog No. 5000295865
Encompass_Preferred

Cholic acid-d5 is the deuterium-labeled analogue of cholic acid supplied for use as an internal standard or tracer in analytical workflows. It supports quantitative method development and routine analyses in NMR, GC-MS, and LC-MS, and is intended for research use only.

  • Deuterium-labeled internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High purity (98.0%).
  • Molecular formula C24H35D5O5; molecular weight 413.60.
  • CAS number 53007-09-3.
  • Provided as a 10 mg vial; store protected from light and under inert atmosphere.
  • For research use only; not for human or animal therapeutic use.

Catalog No. 50-002-95865 Supplier Medchemexpress LLC Supplier No. HYN0324S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cholic acid-d5 is the deuterium-labeled analogue of cholic acid supplied for use as an internal standard or tracer in analytical workflows. It supports quantitative method development and routine analyses in NMR, GC-MS, and LC-MS, and is intended for research use only.

  • Deuterium-labeled internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High purity (98.0%).
  • Molecular formula C24H35D5O5; molecular weight 413.60.
  • CAS number 53007-09-3.
  • Provided as a 10 mg vial; store protected from light and under inert atmosphere.
  • For research use only; not for human or animal therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.