Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cholic acid impurity 1 | 24637-46-5 | 95.1% | 390.6 g/mol | C24H38O4 | 10 MG
SDP

Catalog No. 5000298336
Encompass_Preferred

Cholic acid impurity 1 is a defined impurity of cholic acid provided as a solid reference material for laboratory research. It is supplied in milligram quantities for analytical reference, impurity profiling, and characterization of synthetic intermediates.

  • Offered as a powder suitable for analytical applications.
  • CAS number 24637-46-5.
  • Molecular formula C24H38O4; molecular weight 390.6 g/mol.
  • Typical purity ~95.1%.
  • Packaged in milligram quantities; recommended storage: powder at -20°C, in solution at -80°C.
  • For research use only; not for clinical or human use.

Catalog No. 50-002-98336 Supplier Medchemexpress LLC Supplier No. HY4347510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cholic acid impurity 1 is a defined impurity of cholic acid provided as a solid reference material for laboratory research. It is supplied in milligram quantities for analytical reference, impurity profiling, and characterization of synthetic intermediates.

  • Offered as a powder suitable for analytical applications.
  • CAS number 24637-46-5.
  • Molecular formula C24H38O4; molecular weight 390.6 g/mol.
  • Typical purity ~95.1%.
  • Packaged in milligram quantities; recommended storage: powder at -20°C, in solution at -80°C.
  • For research use only; not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.