Learn More
Cayman Chemical Cholic Acid anilide, Purity: ≥98%, CAS: 6912-96-5, 10 mg
SDP
Catalog No. 502934101
Encompass_Preferred
A synthetic bile acid and derivative of cholic acid; inhibits the germination of C. difficile strain R20291 spores in vitro (IC50 = 1.8 μM)
Catalog No. 50-293-4101 Supplier Cayman Chemical Supplier No. 2614410 MG
A synthetic bile acid and derivative of cholic acid; inhibits the germination of C. difficile strain R20291 spores in vitro (IC50 = 1.8 μM)