Learn More
Abcam Cholic acid, non-denaturing ionic detergent, 50G
Catalog No. 502704741
Encompass_Preferred
MW 408.6 g/mol, Purity >98%. Non-denaturing ionic detergent used for extraction of membrane proteins. Major primary bile acid produced in the liver and usually conjugated with glycine or taurine to form bile salts. Facilitates fat absorption and cholesterol excretion.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4741 Supplier Abcam Supplier No. AB14630450G
MW 408.6 g/mol, Purity >98%. Non-denaturing ionic detergent used for extraction of membrane proteins. Major primary bile acid produced in the liver and usually conjugated with glycine or taurine to form bile salts. Facilitates fat absorption and cholesterol excretion.
The product is subject to the following: Abcam Restricted Use Statement