Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cholic Acid, Sodium Salt, Calbiochem™,

Catalog No. 22910150GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
22-910-150GM 50 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-910-150GM Supplier MilliporeSigma™ Supplier No. 22910150GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 361-09-1
Molecular Formula C24H39NaO5
Molecular Weight (g/mol) 430.56
MDL Number MFCD00150749,MFCD00064138
InChI Key NRHMKIHPTBHXPF-TUJRSCDTSA-M
PubChem CID 57349315
IUPAC Name sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
SMILES [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

Color White
Quantity 50 g
Packaging Glass Bottle
Percent Purity >96% (TLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.