Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholic Acid Sodium Salt, MP Biomedicals™

Catalog No. MP021028973 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021028973 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021028973 Supplier MP Biomedicals, Inc Supplier No. 02102897.3
Add to Cart
Add to Cart

Used in the solubilization of membrane-bound proteins in their native state.

  • A component for the preparation of liposomes.
  • Used in a study to investigate the elastic deformable vesicles, transfersomes, for transdermal drug delivery.
  • Biochemical solubilizing agent (a non-ionic, non-denaturing detergent).
  • Occurs inconjugation with glycine or taurine in bile of most vertebrates .
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 361-09-1
Molecular Formula C24H39NaO5
Molecular Weight (g/mol) 430.56
MDL Number MFCD00150749,MFCD00064138
InChI Key NRHMKIHPTBHXPF-TUJRSCDTSA-M
Synonym sodium cholate, sodium 3,7,12-trihydroxycholan-24-oate, sodium cholate, reagent, ksc575s0b, 3,7,12-trihydroxycholan-24-oic acid sodium salt
PubChem CID 57349315
IUPAC Name sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
SMILES [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

Physical Form White Powder
pH 7.5 to 9.0 (2% aq. soln.)
Quantity 25 g
Solubility Information Soluble in water (100mg/mL). Soluble in ethanol (40mg/mL, clear, colorless solution).
Formula Weight 430.6
Percent Purity ≥98.5%
Concentration or Composition (by Analyte or Components) CriticalMicelle Concentration (CMC) 14mM
Recommended Storage Store at Room Temperature (15°C to 30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.