Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholic acid sodium salt, Thermo Scientific Chemicals

Catalog No. AAJ6205030
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6205030 250 g
AAJ6205014 25 g
AAJ6205022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6205030 Supplier Thermo Scientific Chemicals Supplier No. J6205030
Add to Cart

Biochemical solubilizing agent

Cholic acid sodium salt is a bio-surfactant which can be used as a reducing reagent in aqueous solution. It is also used as a non-denaturing detergent used for extraction of membrane proteins. Sodium cholate hydrate has been used in a study to assess the effect of phospholipid formulation and surfactants on the dermal delivery of hydrophilic drugs from elastic liposomes. It has also been used in a study to investigate the elastic deformable vesicles, transfersomes, for transdermal drug delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cholic acid sodium salt is a bio-surfactant which can be used as a reducing reagent in aqueous solution. It is also used as a non-denaturing detergent used for extraction of membrane proteins. Sodium cholate hydrate has been used in a study to assess the effect of phospholipid formulation and surfactants on the dermal delivery of hydrophilic drugs from elastic liposomes. It has also been used in a study to investigate the elastic deformable vesicles, transfersomes, for transdermal drug delivery.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206986-87-0
Molecular Formula C24H39NaO5
Molecular Weight (g/mol) 430.56
MDL Number MFCD00064138 MFCD00003672
InChI Key NRHMKIHPTBHXPF-TUJRSCDTSA-M
Synonym sodium cholate hydrate, cholalic acid sodium salt, c24h39o5.na.h2o, sodium cholate hydrate, bioxtra, sodium cholate hydrate dried material, nt, sodium cholate hydrate, from ox and/or sheep bile, 3, a,7, a,12, a-trihydroxy-5, a-cholan-24-oic acid sodium salt, sodium cholate hydrate, suitable for cell culture, bioreagent, 4r-4-1s,2s,7s,11s,16s,5r,9r,10r,14r,15r-5,9,16-trihydroxy-2,15-dimethylte tracyclo 8.7.0.0<2,7>.0<11,15> heptadec-14-yl pentanoic acid, sodium salt, hyd rate, sodium cholate hydrate, pharmagrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
PubChem CID 23679061
IUPAC Name sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
SMILES [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Color White
Quantity 250 g
Beilstein 3582354
Merck Index 14,2203
Solubility Information It is soluble in water.
Formula Weight 430.6
Physical Form Powder
Chemical Name or Material Cholic acid sodium salt
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.